Merck
  • Nanonization of curcumin by antisolvent precipitation: process development, characterization, freeze drying and stability performance.
Nanonization of curcumin by antisolvent precipitation: process development, characterization, freeze drying and stability performance.
International journal of pharmaceutics (2014-12-03)
Deepak Yadav, Neeraj Kumar
ABSTRACT
The present work aims to investigate applicability of antisolvent precipitation method for preparation of nanosized curcumin and to control their characteristics by determining the influence of process and solvents on solid-state properties of curcumin nanoparticles. Effects of different experimental parameters on particle size were investigated using dynamic light scattering. Particle morphology was studied using SEM. Drug content in stabilized nanoparticles was determined using HPLC. Residual moisture content after lyophilisation was determined using Karl Fischer method and solid state properties were investigated using DSC, TGA, FTIR and powder-XRD. The resulting product showed a high drug load and contained the drug in amorphous form. The particle diameters of prepared curcumin nanoparticles were found in the range of 100-200 nm. In vitro drug release studies indicated a sustained release profile of curcumin from the nanoparticles. Antisolvent precipitation produced amorphous curcumin nanoparticles whose size and morphology could be controlled using gelatine as stabilizer. Lyophilized curcumin nanoparticles with d-sorbitol as lyoprotectant possessed good redispersibility and showed up to 4 times faster in vitro curcumin release rate than that of unprocessed curcumin. Stability tests (at 2-8°C and ambient conditions) indicated that the product was stable for up to 6 months of storage.
Product Description
Sorbitol, European Pharmacopoeia (EP) Reference Standard
USP
Sorbitol, United States Pharmacopeia (USP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), for molecular biology
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
D-Sorbitol, ≥98% (GC), BioXtra
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Sorbitol, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-Sorbitol, 99% (GC)
Sigma-Aldrich
D-Sorbitol, liquid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbitol F solution, 70 wt. % in H2O, Contains mainly D-sorbitol with lesser amounts of other hydrogenated oligosaccharides
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC