Merck
  • Simvastatin induces the osteogenic differentiation of human periodontal ligament stem cells.
Simvastatin induces the osteogenic differentiation of human periodontal ligament stem cells.
Fundamental & clinical pharmacology (2013-10-12)
Bing-jiao Zhao, Yue-hua Liu
ABSTRACT
Periodontal ligament stem cells (PDLSCs) are considered as potential mesenchymal stem cell sources for future clinical applications in periodontal regeneration therapy. Simvastation, widely used for lowering serum cholesterol, is known to have a bone stimulatory effect. However, it is not clear whether simvastation affects the differentiation of PDLSCs. This study examined the effects of simvastatin on human PDLSCs in vitro and in vivo. Using the limiting dilution technique, human PDLSCs were isolated and expanded. PDLSCs were cultured with simvastatin (0.01-10 μM), and the proliferation was measured. The osteogenic differentiation was characterized by alkaline phosphatase (ALP) activity and Alizarin Red-S staining for calcium deposition. The gene expression levels of osteogenic markers were evaluated by RT-PCR. In addition, PDLSCs were transplanted into nude mice with ceramic bovine bone powders as carriers to observe the capacity of mineralized tissue formation in vivo. Simvastatin at concentrations <1 μM did not suppress the proliferation of PDLSCs. After the administration of 0.1 μM simvastatin, the expression of ALP, bone sialoprotein, and bone morphogenetic protein-2 genes were significantly upregulated, and the ALP activity and mineralized nodule formation were significantly higher in the simvastatin-treated cells than the control cells. In addition, the in vivo transplantation results showed that simvastatin treatment promoted the degree of mineralized tissue formation. Collectively, simvastatin has positive effects on osteogenic differentiation of human PDLSCs in vitro and in vivo. This suggests that simvastatin might be a useful osteogenic induction agent for periodontal bone regeneration.
Product Description
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Alizarin, Dye content 97 %
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, suitable for microscopy
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)