Merck
  • Upregulation of jasmonate biosynthesis and jasmonate-responsive genes in rice leaves in response to a bacterial pathogen mimic.
Upregulation of jasmonate biosynthesis and jasmonate-responsive genes in rice leaves in response to a bacterial pathogen mimic.
Functional & integrative genomics (2014-12-17)
Ashish Ranjan, Jyothilakshmi Vadassery, Hitendra Kumar Patel, Alok Pandey, Ramesh Palaparthi, Axel Mithöfer, Ramesh V Sonti
ABSTRACT
Xanthomonas oryzae pv. oryzae, the causal agent of bacterial blight of rice, secretes several cell wall degrading enzymes including cellulase (ClsA) and lipase/esterase (LipA). Prior treatment of rice leaves with purified cell wall degrading enzymes such as LipA can confer enhanced resistance against subsequent X. oryzae pv. oryzae infection. To understand LipA-induced rice defense responses, microarray analysis was performed 12 h after enzyme treatment of rice leaves. This reveals that 867 (720 upregulated and 147 downregulated) genes are differentially regulated (≥2-fold). A number of genes involved in defense, stress, signal transduction, and catabolic processes were upregulated while a number of genes involved in photosynthesis and anabolic processes were downregulated. The microarray data also suggested upregulation of jasmonic acid (JA) biosynthetic and JA-responsive genes. Estimation of various phytohormones in LipA-treated rice leaves demonstrated a significant increase in the level of JA-Ile (a known active form of JA) while the levels of other phytohormones were not changed significantly with respect to buffer-treated control. This suggests a role for JA-Ile in cell wall damage induced innate immunity. Furthermore, a comparative analysis of ClsA- and LipA-induced rice genes has identified key rice functions that might be involved in elaboration of damage-associated molecular pattern (DAMP)-induced innate immunity.
Product Description
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Coronatine, from Pseudomonas syringae pv Glycinea, >95% (HPLC)
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Salicylic acid, meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Salicylic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Salicylic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Salicylic acid, puriss. p.a., ≥99.0% (T)
Sigma-Aldrich
Salicylic acid, ≥99%, FG
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Salicylic acid, suitable for plant cell culture
Supelco
Salicylic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Salicylic acid, United States Pharmacopeia (USP) Reference Standard