Merck
  • CD200: association with cancer stem cell features and response to chemoradiation in head and neck squamous cell carcinoma.
CD200: association with cancer stem cell features and response to chemoradiation in head and neck squamous cell carcinoma.
Head & neck (2014-04-05)
Yuh-Seog Jung, Paola D Vermeer, Daniel W Vermeer, Sang-Jin Lee, Ah Ra Goh, Hyun-Joo Ahn, John H Lee
ABSTRACT
The purpose of this study was to characterize the expression of CD200, a membrane protein that functions in immune evasion, to examine its correlations with cancer stem cell (CSC)-like features and analyze its response to chemotherapy and radiation in human papillomavirus (HPV)-positive (+) and negative (-) head and neck squamous cell carcinomas (HNSCCs). CD200 expression was analyzed in several HNSCC cell lines. CD200 was overexpressed in HPV(+) murine tonsil epithelial cells, its effects on Shh and Bmi-1 were examined in vitro, and tumor growth and response to chemoradiation were analyzed in vitro and in vivo. CD200 was diversely expressed and consistently associated with expression of Bmi-1 and Shh. Overexpression of CD200 induced Bmi-1 and Shh. Tumors grew similarly between C57BL/6 and Rag1(-/-) C57BL/6 mice. CD200 expression enhanced the resistance to chemoradiation only in vivo. CD200 was related to CSC features and modulates response to chemoradiation in vivo. Attenuating this might be a potential therapeutic strategy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-Bmi-1 Antibody, clone F6, clone F6, Upstate®, from mouse
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
NSC 632839, ≥98% (HPLC)