Merck
  • Identification of a new soybean kunitz trypsin inhibitor mutation and its effect on bowman-birk protease inhibitor content in soybean seed.
Identification of a new soybean kunitz trypsin inhibitor mutation and its effect on bowman-birk protease inhibitor content in soybean seed.
Journal of agricultural and food chemistry (2015-01-23)
Jason D Gillman, Won-Seok Kim, Hari B Krishnan
ABSTRACT
Soybean seed contains antinutritional compounds that inactivate digestive proteases, principally corresponding to two families: Kunitz trypsin inhibitors (KTi) and Bowman-Birk inhibitors (BBI). High levels of raw soybean/soybean meal in feed mixtures can cause poor weight gain and pancreatic abnormalities via inactivation of trypsin/chymotrypsin enzymes. Soybean protein meal is routinely heat-treated to inactivate inhibitors, a practice that is energy-intensive and costly and can degrade certain essential amino acids. In this work, we screened seed from 520 soybean accessions, using a combination of sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and immunoblots with anti-Kunitz trypsin inhibitor antibodies. A soybean germplasm accession was identified with a mutation affecting an isoform annotated as nonfunctional (KTi1), which was determined to be synergistic with a previously identified mutation (KTi3-). We observed significant proteome rebalancing in all KTi mutant lines, resulting in dramatically increased BBI protein levels.
Sigma-Aldrich
N-Benzoyl-L-tyrosine ethyl ester
Sigma-Aldrich
Nα-Benzoyl-L-arginine ethyl ester hydrochloride, trypsin substrate