Merck
  • Prenatal and developmental toxicity study of meclizine and caffeine combination in female albino Wistar rats.
Prenatal and developmental toxicity study of meclizine and caffeine combination in female albino Wistar rats.
Indian journal of experimental biology (2015-02-06)
M Sandeep, Jose M Alvin
ABSTRACT
Meclizine and caffeine combination is used for the treatment of morning sickness. Both compounds are teratogenic and caffeine is known to possess anti-fertility activity also. The present study was undertaken to evaluate the reproductive toxic effect of meclizine and caffeine combination. Three doses were taken for the study; low dose (LD; meclizine 3.7 mg/kg and caffeine 3 mg/kg) was selected from commercially available formulation, middle dose (MD; meclizine 37 mg/kg and caffeine 30 mg/kg) and high dose (HD; meclizine 370 mg/kg and caffeine 300 mg/kg). The mixture was administered 1-7 days and 8-14 days for fertility and embryotoxic studies respectively. Laparotomy was done on 10t day of gestation period. Number of implants and corpora lutea were counted, pre and post-implantation losses were determined. In embryo toxicity study fetuses were evaluated for external, skeletal and visceral examination. High dose was removed from both fertility and embryotoxicity studies due to its severe toxicity to the dam. Significant anti-fertility activity was observed at middle dose. Embryotoxicity study showed significant reduction in fetal body weight, body length and body mass index, dam body weight gain on gestation day 14. Absolute kidney weight in MD and absolute and relative spleen weight in both LD and MD were significantly reduced. There was no increase in external or internal congenital anomalies at both LD and MD. The, results suggest that prescription of meclizine and caffeine for morning sickness in early pregnancy should be reviewed carefully.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Caffeine, European Pharmacopoeia (EP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard