Merck
  • Genetic manipulation of the ghrelin signaling system in male mice reveals bone compartment specificity of acylated and unacylated ghrelin in the regulation of bone remodeling.
Genetic manipulation of the ghrelin signaling system in male mice reveals bone compartment specificity of acylated and unacylated ghrelin in the regulation of bone remodeling.
Endocrinology (2014-07-26)
Patric J D Delhanty, Martijn van der Velde, Bram C J van der Eerden, Yuxiang Sun, Julia M M Geminn, Aart-Jan van der Lely, Roy G Smith, Johannes P T M van Leeuwen
ABSTRACT
Ghrelin receptor-deficient (Ghsr-/-) mice that lack acylated ghrelin (AG) signaling retain a metabolic response to unacylated ghrelin (UAG). Recently, we showed that Ghsr-deficiency affects bone metabolism. The aim of this study was to further establish the impact of AG and UAG on bone metabolism. We compared bone metabolism in Ghsr-/- (lacking only AG signaling) and ghrelin-deficient (Ghrl-/-; both AG and UAG deficient) male mice. Ghrl-/- mice had lower cortical bone mass, whereas Ghsr-/- mice had lower trabecular bone mass. This demonstrates bone compartment-specific effects of AG and a role for UAG in bone metabolism. Also, Ghrl-/- but not Ghsr-/- mice had increased bone formation rate and increased osteogenic stem cell numbers in their bone marrow. In ex vivo bone marrow cultures both AG and UAG inhibited osteoblast differentiation. This indicated that bone resorption must be increased in these mice. Accordingly, osteoclastogenesis rate was faster in bone marrow cultures from Ghsr-/- and Ghrl-/- mice, and osteoclast formation was inhibited by AG signaling and partially suppressed by UAG. In osteoblast cultures, AG markedly induced osteoprotegerin gene expression and both peptides reduced RANKL/osteoprotegerin ratio. These data describe unique cell-type specific effects of AG and UAG within a single tissue, supporting a tight and complex control of bone formation and resorption as well as a link between nutrition and bone metabolism. The balance between AG and UAG actions in the bone marrow may lead to bone compartmental-specific effects.
Product Description
Methyl methacrylate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Methyl methacrylate, contains ≤30 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Methyl methacrylate, 99%, stabilized
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Amphotericin B, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Hematoxylin, certified by the Biological Stain Commission
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hematoxylin
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard