Merck
  • Derivatization method for sensitive determination of 3-hydroxybenzo[a]pyrene in human urine by liquid chromatography-electrospray tandem mass spectrometry.
Derivatization method for sensitive determination of 3-hydroxybenzo[a]pyrene in human urine by liquid chromatography-electrospray tandem mass spectrometry.
Journal of chromatography. A (2015-01-04)
Kai Luo, Qun Gao, Jianying Hu
ABSTRACT
While urinary 3-hydroxybenzo[a]pyrene (3-OHBaP) can indicate exposure to carcinogenic benzo[a]pyrene (BaP) in humans, most previous analytical methods could not detect it in the general population due to a lack of sufficient sensitivity. In this study, a new liquid chromatography-electrospray mass-mass spectrometry (LC-ESI-MS/MS) method was developed to analyze 3-OHBaP by employing an optimized derivatization reaction with dansyl chloride (DNS) in the aqueous phase. The instrument detection limit (IDL) of the newly developed method was 10ng/L, which is 60-fold lower than without derivatization. The method was successfully applied to analyze 3-OHBaP and 1-hydroxypyrene (1-OHPy) in human urine combined with enzymatic hydrolysis, C18 enrichment, liquid-liquid extraction and silica cartridge cleanup. The overall method recoveries were 68.7±5.47% for 3-OHBaP and 72.3±4.96% for 1-OHPy, with matrix effects of <10%. The limits of quantification (LOQs) in urine were 0.3±0.02ng/L and 0.5±0.02ng/L (n=3), respectively. The method was applied to analyze seven urine samples of volunteers, and 3-OHBaP was detected in five samples. The concentration of urinary 3-OHBaP ranged from <LOD to 2.30ng/L, much lower than those (42.0-216ng/L) of 1-OHPy. The developed method provides a new way to sensitively determine 3-OHBaP in urine of the general population.
Product Description
USP
Valacyclovir Related Compound G, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triethylamine, for protein sequence analysis, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%