Merck
  • Effects of age and smoking on endothelial function assessed by quantitative cardiovascular magnetic resonance in the peripheral and central vasculature.
Effects of age and smoking on endothelial function assessed by quantitative cardiovascular magnetic resonance in the peripheral and central vasculature.
Journal of cardiovascular magnetic resonance : official journal of the Society for Cardiovascular Magnetic Resonance (2015-04-18)
Michael C Langham, Yongxia Zhou, Erica N Chirico, Jeremy F Magland, Chandra M Sehgal, Erin K Englund, Emile R Mohler, Wensheng Guo, Suliman Barhoum, Felix W Wehrli
ABSTRACT
Both age and smoking promote endothelial dysfunction and impair vascular reactivity. Here, we tested this hypothesis by quantifying new cardiovascular magnetic resonance (CMR)-based biomarkers in smokers and nonsmokers. Study population: young non-smokers (YNS: N = 45, mean age = 30.2 ± 0.7 years), young smokers (YS: N = 39 mean age 32.1 ± 0.7 years), older non-smokers (ONS: N = 45, mean age = 57.8 ± 0.6 years), and older smokers (OS: N = 40, mean age = 56.3 ± 0.6 years), all without overt cardiovascular disease. Vascular reactivity was evaluated following cuff-induced hyperemia via time-resolved blood flow velocity and oxygenation (SvO2) in the femoral artery and vein, respectively. SvO2 dynamics yielded washout time (time to minimum SvO2), resaturation rate (upslope) and maximum change from baseline (overshoot). Arterial parameters included pulse ratio (PR), hyperemic index (HI) and duration of hyperemia (TFF). Pulse-wave velocity (PWV) was assessed in aortic arch, thoracoabdominal aorta and iliofemoral arteries. Ultrasound-based carotid intimal-medial thickness (IMT) and brachial flow-mediated dilation were measured for comparison. Age and smoking status were independent for all parameters. Smokers had reduced upslope (-28.4%, P < 0.001), increased washout time (+15.3%, P < 0.01), and reduced HI (-19.5%, P < 0.01). Among non-smokers, older subjects had lower upslope (-22.7%, P < 0.01) and overshoot (-29.4%, P < 0.01), elevated baseline pulse ratio (+14.9%, P < 0.01), central and peripheral PWV (all P < 0.05). Relative to YNS, YS had lower upslope (-23.6%, P < 0.01) and longer washout time (13.5%, P < 0.05). Relative to ONS, OS had lower upslope (-33.0%, P < 0.01). IMT was greater in ONS than in YNS (+45.6%, P < 0.001), and also in YS compared to YNS (+14.7%, P < 0.05). Results suggest CMR biomarkers of endothelial function to be sensitive to age and smoking independent of each other.
Sigma-Aldrich
Hydriodic acid, 57 wt. %, distilled, 99.999% trace metals basis
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, ACS reagent, 55%
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%
Sigma-Aldrich
Hydriodic acid, 57 wt. % in H2O, distilled, stabilized, 99.95%
Sigma-Aldrich
Hydriodic acid, contains no stabilizer, distilled, 57 wt. % in H2O, 99.99% trace metals basis