Merck
  • Anti-inflammatory effects of atorvastatin treatment in chronic obstructive pulmonary disease. A controlled pilot study.
Anti-inflammatory effects of atorvastatin treatment in chronic obstructive pulmonary disease. A controlled pilot study.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society (2015-02-27)
R M Mroz, P Lisowski, A Tycinska, J Bierla, P Z Trzeciak, L Minarowski, R Milewski, A Lisowska, P Boros, B Sobkowicz, A M Duszewska, E Chyczewska, W J Musial, W MacNee
ABSTRACT
Observational studies have suggested that statins may have beneficial effects on outcomes in chronic obstructive pulmonary disease (COPD) patients. These effects may be mediated through an anti-inflammatory effect of statins. The purpose of this pilot-study was to determine whether statins have an anti-inflammatory effect on the lungs of COPD patients. We conducted randomized, controlled, parallel group pilot-study to compare the effects of atorvastatin (n=12) or placebo (n=6) on lung inflammation in patients with mild to moderate COPD. The primary endpoint was change in CD45+ cells expression measured by immunohistochemistry and changes in expression of genes measured using microarrays in lung biopsy (TBB) samples before and after 12 weeks of treatment with atorvastatin 40 mg/day. All subjects had spirometry, lung volumes, diffusing capacity of the lungs for carbon monoxide (DLCO), St George's Respiratory Questionnaire (SGRQ), 6 minute walk distance (6 MWD), serum lipids, hs-CRP, induced sputum (IS), bronchoscopy and TBB carried out at baseline and after treatment. TBB specimens were processed for histology, immunohistochemistry and genome-wide association studies (GWAS) profiling. Seventeen subjects completed the study. There was a significant improvement in SGRQ with mean SGRQ decreased by 12 points after treatment with atorvastatin (P=0.012). Atorvastatin treatment produced a significant 34% reduction in sputum neutrophil count, and a 57% reduction in CD45+ cells in lung biopsies (expressed as integrated optical density -IOD; median IOD 62.51% before, 27.01% after atorvastatin treatment, P=0.008). In patients' lung tissue atorvastatin treatment produced downregulation of key genes involved in inflammatory processes, immune response, and leukocyte activation. These data demonstrate the pulmonary anti-inflammatory effects of atorvastatin in COPD patients with the potential for beneficial clinical effects. ClinicalTrials.gov: NCT01748279.
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Creatine, anhydrous