Merck
  • A separation of tyramine on a 2-(4-methoxyphenyl)ethylamine imprinted polymer: an answer from theoretical and experimental studies.
A separation of tyramine on a 2-(4-methoxyphenyl)ethylamine imprinted polymer: an answer from theoretical and experimental studies.
Talanta (2014-08-17)
Piotr Luliński, Monika Sobiech, Teresa Zołek, Dorota Maciejewska
ABSTRACT
A 2-(4-methoxyphenyl)ethylamine imprinted polymer (MIP) was successfully applied for the selective separation of tyramine. A computational analysis was used to predict the affinity of the polymer matrix towards tyramine and a preliminary experimental evaluation was made for the target analyte. Then the experimental analysis of polymer towards tyramine was continued. The binding sites were characterized with employment of the Langmuir and Freudlich models. After the optimization of solid phase extraction towards tyramine, the most appropriate systems for the extraction steps were chosen: methanol-water 85:15 v/v for the loading and the washing as well as 0.04 M aq. ammonium acetate-methanol 30:70 v/v for the elution steps. The biogenic compounds as tryptamine, serotonin, octopamine, synephrine, and l-tyrosine were used for the selectivity study on the basis of binding capacities of the analytes on the imprinted and the non-imprinted polymers. The theoretical approach to obtained results allowed to explain the adsorption selectivity of the tested polymer. Finally, the complex matrix of bovine serum albumin was used to show the usefulness of imprinted material for bioanalysis. The obtained recoveries showed the superiority of MIP over the commercial sorbent C18. Total recoveries of tyramine from spiked bovine serum albumin sample were determined as: 95±2%, 14±3%, and 1.9±0.4% for the imprinted, non-imprinted, and commercial C18 sorbents, respectively.
Sigma-Aldrich
L-Tyrosine, FG
Supelco
Acetone, analytical standard
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
L-Tyrosine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tyrosine, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
SAFC
L-Tyrosine
Supelco
L-Tyrosine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tyrosine, BioUltra, ≥99.0% (NT)
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
L-Tyrosine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formamide, spectrophotometric grade, ≥99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Formamide, ReagentPlus®, ≥99.0% (GC)
Supelco
L-Tyrosine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%