Merck
  • Enhancement and induction of HIV-1 infection through an assembled peptide derived from the CD4 binding site of gp120.
Enhancement and induction of HIV-1 infection through an assembled peptide derived from the CD4 binding site of gp120.
Chembiochem : a European journal of chemical biology (2015-02-03)
Andrea Groß, Katja Rödel, Barbara Kneidl, Norbert Donhauser, Marek Mössl, Edina Lump, Jan Münch, Barbara Schmidt, Jutta Eichler
ABSTRACT
Contact between the human immunodeficiency virus (HIV-1) and its target cell is initiated by the interaction of viral gp120 with cellular CD4. An assembled peptide (CD4bs-M) that presents the CD4 binding site of gp120 was previously shown to inhibit the gp120-CD4 interaction. Here, we demonstrate that CD4bs-M selectively enhances infection of cells with HIV-1, whereas infection with herpes simplex virus remains largely unaffected. The effects of CD4bs-M variants containing D-amino acids, or prolines at selected positions, point to the importance of side chain orientation and spatial orientation of this fragment. Furthermore, CD4bs-M was shown to assemble into amyloid-like fibrils that capture HIV-1 particles, which likely contributes to the infection-enhancing effect. Beyond infection enhancement, CD4bs-M enabled HIV-1 infection of CD4-negative cells, suggesting that binding of the peptide to gp120 facilitates interaction of gp120 with coreceptors, which might in turn enhance HIV-1 entry.
Product Description
Supelco
Density Standard 998 kg/m3, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water, BioPerformance Certified
Supelco
Water, ACS reagent, for ultratrace analysis
Supelco
Water, for TOC analysis
Supelco
Water, suitable for ion chromatography
Sigma-Aldrich
Water, tested according to Ph. Eur.
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, absolute, over molecular sieve (H2O ≤0.04%), ≥99.5% (GC)
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Supelco
Water, for HPCE, for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, reagent grade
Sigma-Aldrich
Water, deuterium-depleted, ≤1 ppm (Deuterium oxide)
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Water, for molecular biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, suitable for HPLC
Sigma-Aldrich
Water, Deionized
Sigma-Aldrich
1,3-Dimethyl-2-imidazolidinone, ≥99.0% (GC)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Pure Water Density Standard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Water, ACS reagent
Sigma-Aldrich
6-Phosphonohexanoic acid, 97%
Supelco
Trifluoroacetic acid, analytical standard
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%