Merck
  • Simultaneous derivatization and solid-based disperser liquid-liquid microextraction for extraction and preconcentration of some antidepressants and an antiarrhythmic agent in urine and plasma samples followed by GC-FID.
Simultaneous derivatization and solid-based disperser liquid-liquid microextraction for extraction and preconcentration of some antidepressants and an antiarrhythmic agent in urine and plasma samples followed by GC-FID.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-01-27)
Mir Ali Farajzadeh, Parisa Khorram, Houshang Ghorbanpour
ABSTRACT
The present work is based on a one-step method including derivatization and solid-based disperser liquid-liquid microextraction followed by gas chromatography-flame ionization detection (GC-FID) for the determination of four antidepressants (fluoxetine, fluvoxamine, tranylcypromine, and nortriptyline) and an antiarrhythmic agent (mexiletine) in human urine and plasma samples. In this method, a mixture of 1,1,2,2-tetrachloroethane (extraction solvent) and butylchloroformate (derivatizing reagent) is added on a sugar cube (solid disperser) and it is introduced into an aqueous sample containing the analytes and a catalyst, e.g. 3-methylpyridine (picoline). During dissolving the sugar cube by manual shaking, the extractant and derivatization agent are gradually released into the sample as very fine droplets. Then the resulted cloudy solution is centrifuged and the sedimented phase is analyzed by GC-FID. The influence of several variables on the efficiency of derivatization/microextraction procedure such as kind and volume of extraction solvent, type and amount of disperser, amount of derivatization agent, and catalyst volume are optimized. Under the optimum conditions the calibration curves are linear in the range of 8-100,000μgL(-1) (coefficient of determination ≥0.994). The relative standard deviations are obtained in the range of 3.0-6.0% for all compounds. Moreover, the detection limits and enrichment factors of the target analytes are obtained in the ranges 1-15μgL(-1) and 228-268, respectively, for both plasma and urine samples. The relative recoveries obtained for the spiked plasma and urine samples are between 70 and 91%. The results show that the proposed method is simple, reliable, low cost, and applicable to determine trace amounts of the studied drugs in biological samples.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Supelco
Carbon tetrachloride, analytical standard
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
1,1,2,2-Tetrachloroethane, analytical standard
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Carbon tetrachloride, anhydrous, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material