Merck
  • Tumor progression is associated with increasing CD11b+ cells and CCL2 in Lewis rat sarcoma.
Tumor progression is associated with increasing CD11b+ cells and CCL2 in Lewis rat sarcoma.
Anticancer research (2015-02-11)
Rajbardhan Mishra, Jana Kovalska, Jozef Janda, Luca Vannucci, Radko Rajmon, Vratislav Horak
ABSTRACT
Tumor models are essential for basic anticancer research and development of novel therapies. In this study, we used a rat sarcoma model in which subcutaneous tumor develops after D6 cell inoculation. The aim of the current study was to analyze changes in haematological parameters, immune cell sub-populations and cytokine profiling during tumor growth, after tumor excision and after second inoculation of D6 cells. Tumor progression was found to be associated with an increased number of leukocytes and increased proportion of CD11b+ cells in peripheral blood. Serum concentration of chemokine (c-c motif) ligand 2, L-selectin and intra cellular adhesion molecule-1 also increased with growing tumor. However, the proportion of CD4+, CD8+ and MHC II+ cells decreased with growth of tumors. After tumor excision, all these parameters returned to pre-inoculation levels and did not change even after a second inoculation of D6 cells. Moreover, absence of secondary tumors after second inoculation of D6 cells gives an insight into development of antitumor immunity stimulated by primary tumor.
Product Description
SAFC
HEPES
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
USP
Propyl gallate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, powder
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material