Merck
  • Relationships between degree of polymerization and antioxidant activities: a study on proanthocyanidins from the leaves of a medicinal mangrove plant Ceriops tagal.
Relationships between degree of polymerization and antioxidant activities: a study on proanthocyanidins from the leaves of a medicinal mangrove plant Ceriops tagal.
PloS one (2014-10-15)
Hai-Chao Zhou, Nora Fung-yee Tam, Yi-Ming Lin, Zhen-Hua Ding, Wei-Ming Chai, Shu-Dong Wei
ABSTRACT
Tannins from the leaves of a medicinal mangrove plant, Ceriops tagal, were purified and fractionated on Sephadex LH-20 columns. 13C nuclear magnetic resonance (13C-NMR), reversed/normal high performance liquid chromatography electrospray ionization mass spectrometry (HPLC-ESI MS) and matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDT-TOF MS) analysis showed that the tannins were predominantly B-type procyanidins with minor A-type linkages, galloyl and glucosyl substitutions, and a degree of polymerization (DP) up to 33. Thirteen subfractions of the procyanidins were successfully obtained by a modified fractionation method, and their antioxidant activities were investigated using 2,2-diphenyl-1-picrylhydrazyl (DPPH) scavenging capacity and ferric reducing antioxidant power (FRAP) method. All these subfractions exhibited potent antioxidant activities, and eleven of them showed significantly different mean DP (mDP) ranging from 1.43±0.04 to 31.77±1.15. Regression analysis demonstrated that antioxidant activities were positively correlative with mDP when around mDP <10, while dropped and then remained at a level similar to mDP = 5 with around 95 µg ml(-1) for DPPH scavenging activity and 4 mmol AAE g(-1) for FRAP value.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzyl mercaptan, 99%, FG
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, >99.0% (HPLC)
Sigma-Aldrich
Benzyl mercaptan, 99%
Sigma-Aldrich
Cesium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, ≥98%
Sigma-Aldrich
Cesium chloride, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
2,5-Dihydroxybenzoic acid, ≥99.5% (HPLC), Ultra pure
Sigma-Aldrich
Cesium chloride, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, ReagentPlus®, 99.9%
Sigma-Aldrich
Cesium chloride, puriss. p.a., ≥99.5%
Supelco
2,5-Dihydroxybenzoic acid, analytical standard
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, 99%
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. of Fe, ≥99.0% (HPLC)