Merck
  • Modulation of hepatitis C virus core DNA vaccine immune responses by co-immunization with CC-chemokine ligand 20 (CCL20) gene as immunoadjuvant.
Modulation of hepatitis C virus core DNA vaccine immune responses by co-immunization with CC-chemokine ligand 20 (CCL20) gene as immunoadjuvant.
Molecular biology reports (2014-06-29)
Christine Hartoonian, Zargham Sepehrizadeh, Mehdi Mahdavi, Arash Arashkia, Yon Suk Jang, Maasoumeh Ebtekar, Mojtaba Tabatabai Yazdi, Babak Negahdari, Azita Nikoo, Kayhan Azadmanesh
ABSTRACT
Plasmid DNA vaccination is a promising vaccine platform for prevention and treatment of infectious disease. Enhancement of the DNA vaccine potency by co-inoculation of immunoadjuvant has been shown to be an effective strategy. Modulation of dendritic cells and T-cells locomotion and trafficking to prime an immune response is mediated by distinct chemokines. The recent study was designed to elucidate the adjuvant activity of plasmid expressing CC-chemokine ligand 20 (pCCL20) in co-inoculation with hepatitis C virus (HCV) core DNA vaccine immunization. pCCL20 was constructed and evaluated for its functional expression. Sub-cutaneous inoculation of pCCL20 with HCV core DNA vaccine was performed via electroporation in BALB/c mice on day 0 and 14 and a HCV core protein booster was applied on day 28. On week after final immunization, both humoral and cell-mediated immune responses were assessed by indirect ELISA for core specific antibodies, lymphocyte proliferation, cytokine ELISA/ELISpot and cytotoxic Grenzyme B (GrzB) release assays. Mice were co-immunized with pCCL20 developed higher levels of core specific IFN-γ/IL-4 ratio and IL-2 release, IFN-γ producing cells, lymphocyte proliferation and cytotoxic Grenzyme B release in both draining lymph nodes and spleen cells of immunized mice. The core-specific serum total IgG and IgG2a/IgG1 ratio were significantly higher when the pCCL20 was co-inoculated. These results suggest the potential of CCL20 chemokine as vaccine adjuvant to enhance Th1 mediated cellular and humoral immune responses in HCV core DNA immunization.
Sigma-Aldrich
Rat Ccl20 / C-C Motif Chemokine 20 ELISA Kit
Sigma-Aldrich
Cotton Rat IL4 / Interleukin 4 ELISA Kit
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Mouse IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IL-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
Rat IL-4 ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Rat IL-4 ELISA Kit, for serum, plasma and cell culture supernatant