Merck
  • Chemical constituents and biological activities of Cirsium leucopsis, C. sipyleum, and C. eriophorum.
Chemical constituents and biological activities of Cirsium leucopsis, C. sipyleum, and C. eriophorum.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2015-02-26)
Mehmet Boğa, Pelin Köseoğlu Yilmaz, Deniz Bariş Cebe, Mashhad Fatima, Bina S Siddiqui, Ufuk Kolak
ABSTRACT
Two endemic Cirsium species, C. leucopsis DC. and C. sipyleum O. Schwarz, and C. eriophorum (L.) Scop. growing in Turkey were investigated to establish their secondary metabolites, fatty acid compositions, and antioxidant and anticholinesterase potentials. Spectroscopic methods were used to elucidate the structures of thirteen known compounds (p-hydroxy-benzoic acid, vanillic acid, cis-epoxyconiferyl alcohol, syringin, balanophonin, 1'-O-methyl-balanophonin, apigenin, kaempferol-3- O-β-D-glucopyranoside, kaempferol-3-O-α-L-rhamnopyranoside, taraxasterol, taraxasterol acetate, β-sitosterol, β-sitosterol-3-O-β-D-glucopyranoside). cis-Epoxyconiferyl alcohol and 1'-O-methyl- balanophonin were isolated for the first time from Cirsium species. Palmitic acid (47.1%) was found to be the main fatty acid of C. leucopsis, linoleic acid in both C. sipyleum (42.1%) and C. eriophorum (37.8 %). Assays of β-carotene bleaching, scavenging of 1,1-diphenyl-2-picrylhydrazyl (DPPH) free radicals, 2,2-azinobis (3-ethylbenzothiazoline-6-sulfonic acid) diammonium (ABTS) cation radicals, and superoxide anion radicals, as well as cupric reducing antioxidant capacity (CUPRAC) were used to determine the antioxidant activities of the extracts and isolated compounds. Vanillic acid, balanohonin, and kaempferol-3-O-aαL-rhamnopyranoside exhibited strong antioxidant activity. Taraxa-terol was a potent inhibitor of acetyl- and butyrylcholinesterase activity, respectively.
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Supelco
Guaiacol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Supelco
Butylated Hydroxytoluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
USP
Butylated hydroxytoluene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Neocuproine, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Potassium peroxodisulfate, puriss. p.a., ACS reagent, ≥99.0% (RT)
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Supelco
Linoleic acid, analytical standard
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
1,3-Propylene sulfite, 99%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer