Merck
  • Carbamylation of immunoglobulin abrogates activation of the classical complement pathway.
Carbamylation of immunoglobulin abrogates activation of the classical complement pathway.
European journal of immunology (2014-08-19)
Catalin Koro, Ewa Bielecka, Anders Dahl-Knudsen, Jan J Enghild, Carsten Scavenius, Johan G Brun, Veronika Binder, Annelie Hellvard, Brith Bergum, Roland Jonsson, Jan Potempa, Anna M Blom, Piotr Mydel
ABSTRACT
Post-translational modifications of proteins significantly affect their structure and function. The carbamylation of positively charged lysine residues to form neutral homoitrulline occurs primarily under inflammatory conditions through myeloperoxidase-dependent cyanate (CNO-) formation. We analyzed the pattern of human IgG1 carbamylation under inflammatory conditions and the effects that this modification has on the ability of antibodies to trigger complement activation via the classical pathway. We found that the lysine residues of IgG1 are rapidly modified after brief exposure to CNO- . Interestingly, modifications were not random, but instead limited to only few lysines within the hinge area and the N-terminal fragment of the CH2 domain. A complement activation assay combined with mass spectrometry analysis revealed a highly significant inverse correlation between carbamylation of several key lysine residues within the hinge region and N-terminus of the CH2 domain and the proper binding of C1q to human IgG1 followed by subsequent complement activation. This severely hindered complement-dependent cytotoxicity of therapeutic IgG1 . The reaction can apparently occur in vivo, as we found carbamylated antibodies in synovial fluid from rheumatoid arthritis patients. Taken together, our data suggest that carbamylation has a profound impact on the complement-activating ability of IgG1 and reveals a pivotal role for previously uncharacterized lysine residues in this process.
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
SAFC
Iodoacetamide
Sigma-Aldrich
Cadmium sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Supelco
Sulfuric acid concentrate, 0.1 M H2SO4 in water (0.2N), eluent concentrate for IC
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Thiosemicarbazide, puriss. p.a., 98%
Supelco
2,3-Butanedione monoxime, for spectrophotometric det. of urea, ≥99.0%
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Thiosemicarbazide, 99%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
2,3-Butanedione monoxime, ≥98%
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.