Merck
  • The effects of screw configuration and polymeric carriers on hot-melt extruded taste-masked formulations incorporated into orally disintegrating tablets.
The effects of screw configuration and polymeric carriers on hot-melt extruded taste-masked formulations incorporated into orally disintegrating tablets.
Journal of pharmaceutical sciences (2014-11-21)
Joseph T Morott, Manjeet Pimparade, Jun-Bom Park, Chelsea P Worley, Soumyajit Majumdar, Zhuoyang Lian, Elanor Pinto, Yunxia Bi, Thomas Durig, Michael A Repka
ABSTRACT
The primary aim of this research was to produce successfully taste masked formulations of Sildenafil Citrate (SC) using hot-melt extrusion (HME) technology. Multiple screw configurations and polymeric carriers were evaluated for their effects on taste masking efficiency, which was assessed by both E-tongue analysis and in vitro dissolution in simulated salivary fluid (SSF, pH 6.8 artificial saliva). The screw configurations were further assessed for their effects on the morphology of the API using PXRD, FT-IR and mid-infrared chemical imaging. It was determined that the screw configuration had a profound effect on the taste masking efficiency of the formulations as a result of altering the physical state of the API. Selected extruded formulations using ethylcellulose (EC) with a pore former were further formulated into orally disintegrating tablets (ODTs), which were optimized by varying the grade and percentage of the superdisintegrant used. An optimized disintegration time of approximately 8 seconds was achieved. The final ODT formulation exhibited excellent taste masking properties with over 85% drug release in gastric media as well as physical tablet properties. Interestingly, friability, which tends to be a common concern when formulating ODTs, was well within the acceptable limits (<1%) for common tablets.
Sigma-Aldrich
Butylated hydroxytoluene, ≥99%, FCC, FG
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Calcium chloride dihydrate, tested according to Ph. Eur.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Magnesium oxide, BioUltra, ≥97.0% (calcined substance, KT)
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Magnesium oxide, tested according to Ph. Eur., heavy
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Magnesium oxide, nanopowder, ≤50 nm particle size (BET)
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium oxide, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Magnesium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride dihydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Calcium chloride dihydrate, for molecular biology, ≥99.0%
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M