Merck
  • The urinary proteome and metabonome differ from normal in adults with mitochondrial disease.
The urinary proteome and metabonome differ from normal in adults with mitochondrial disease.
Kidney international (2014-09-11)
Andrew M Hall, Annalisa Vilasi, Isabel Garcia-Perez, Marta Lapsley, Charlotte L Alston, Robert D S Pitceathly, Robert McFarland, Andrew M Schaefer, Doug M Turnbull, Nick J Beaumont, Justin J Hsuan, Pedro R Cutillas, John C Lindon, Elaine Holmes, Robert J Unwin, Robert W Taylor, Grainne S Gorman, Shamima Rahman, Michael G Hanna
ABSTRACT
We studied the extent and nature of renal involvement in a cohort of 117 adult patients with mitochondrial disease, by measuring urinary retinol-binding protein (RBP) and albumin; established markers of tubular and glomerular dysfunction, respectively. Seventy-five patients had the m.3243A>G mutation and the most frequent phenotypes within the entire cohort were 14 with MELAS, 33 with MIDD, and 17 with MERRF. Urinary RBP was increased in 29 of 75 of m.3243A>G patients, whereas albumin was increased in 23 of the 75. The corresponding numbers were 16 and 14, respectively, in the 42 non-m.3243A>G patients. RBP and albumin were higher in diabetic m.3243A>G patients than in nondiabetics, but there were no significant differences across the three major clinical phenotypes. The urine proteome (mass spectrometry) and metabonome (nuclear magnetic resonance) in a subset of the m.3243A>G patients were markedly different from controls, with the most significant alterations occurring in lysosomal proteins, calcium-binding proteins, and antioxidant defenses. Differences were also found between asymptomatic m.3243A>G carriers and controls. No patients had an elevated serum creatinine level, but 14% had hyponatremia, 10% had hypophosphatemia, and 14% had hypomagnesemia. Thus, abnormalities in kidney function are common in adults with mitochondrial disease, exist in the absence of elevated serum creatinine, and are not solely explained by diabetes.
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Methanol, NMR reference standard
Millipore
Urea solution, suitable for microbiology, 40% in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Urea, BioUltra, for molecular biology, 99% (T)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%