Merck
  • The development and validation of a rapid method for the determination of antimicrobial agent residues in milk and meat using ultra performance liquid chromatography coupled to quadrupole--Orbitrap mass spectrometry.
The development and validation of a rapid method for the determination of antimicrobial agent residues in milk and meat using ultra performance liquid chromatography coupled to quadrupole--Orbitrap mass spectrometry.
Journal of pharmaceutical and biomedical analysis (2014-10-06)
Guntis Cepurnieks, Jekaterina Rjabova, Dzintars Zacs, Vadims Bartkevics
ABSTRACT
A new multi-class method has been developed for the identification and quantification of the residues of 26 antibiotics from different classes (sulfonamides, macrolides, tetracyclines, penicillins, and quinolones) in milk and meat by ultra performance liquid chromatography coupled to hybrid quadrupole - high resolution Orbitrap mass spectrometry (UPLC-qOrbitrap). The sample preparation included extraction of two analytical portions with acetonitrile and 5% trichloroacetic acid, respectively, followed by centrifugation and filtration. The method was validated over three days at 50% of MRL (maximum residue limit) set in the European Union. Experiments on spiked meat and milk samples showed that the average recovery of the antibiotics ranged from 83% to 112%, and the coefficients of variation were between 8.9% and 39%.
Product Description
Supelco
Sulfamethazine, VETRANAL®, analytical standard
Supelco
Sulfadimethoxine, VETRANAL®, analytical standard
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Supelco
Sulfadimethoxine, 98.0-102.0%
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sulfamethazine, 99.0-101.0% (on dried basis)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Millipore
Chlortetracycline Selective Supplement, suitable for microbiology
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Supelco
Marbofloxacin, VETRANAL®, analytical standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, NMR reference standard
Marbofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Oxytetracycline, Pharmaceutical Secondary Standard; Certified Reference Material
Sulfadimethoxine for peak identification, European Pharmacopoeia (EP) Reference Standard
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sulfadimethoxine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Enrofloxacin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%