Merck
  • Branched amphiphilic cationic oligopeptides form peptiplexes with DNA: a study of their biophysical properties and transfection efficiency.
Branched amphiphilic cationic oligopeptides form peptiplexes with DNA: a study of their biophysical properties and transfection efficiency.
Molecular pharmaceutics (2015-02-04)
L Adriana Avila, Luana R M M Aps, Pinakin Sukthankar, Nicoleta Ploscariu, Sushanth Gudlur, Ladislav Šimo, Robert Szoszkiewicz, Yoonseong Park, Stella Y Lee, Takeo Iwamoto, Luis C S Ferreira, John M Tomich
ABSTRACT
Over the past decade, peptides have emerged as a new family of potential carriers in gene therapy. Peptides are easy to synthesize and quite stable. Additionally, sequences shared by the host proteome are not expected to be immunogenic or trigger inflammatory responses, which are commonly observed with viral approaches. We recently reported on a new class of branched amphiphilic peptide capsules (BAPCs) that self-assemble into extremely stable nanospheres. These capsules are capable of retaining and delivering alpha-emitting radionuclides to cells. Here we report that, in the presence of double stranded plasmid DNA, BAPCs are unable to form. Instead, depending of the peptide/DNA ratios, the peptides either coat the plasmid surface forming nanofibers (high peptide to DNA ratio) or condense the plasmid into nanometer-sized compacted structures (at low peptide to DNA ratios). Different gene delivery efficiencies are observed for the two types of assemblies. The compacted nanometer-sized structures display much higher transfection efficiencies in HeLa cells. This level of transfection is greater than that observed for a lipid-based reagent when the total number of viable transfected cells is taken into account.
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Trypan Blue solution, 0.4%, for microscopy
Sigma-Aldrich
Glycerin, meets USP testing specifications
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)