Merck
  • PCB153, TCDD and estradiol compromise the benzo[a]pyrene-induced p53-response via FoxO3a.
PCB153, TCDD and estradiol compromise the benzo[a]pyrene-induced p53-response via FoxO3a.
Chemico-biological interactions (2014-06-24)
Lauy Al-Anati, Sandeep Kadekar, Johan Högberg, Ulla Stenius
ABSTRACT
TCDD, polychlorinated biphenyls (PCB) and polycyclic aromatic hydrocarbons (PAH) coexist in the environment. However, there are few studies on combined effects of these compounds. We have studied the effect of TCDD, PCB153 and estradiol on p53 signaling induced by PAHs. We show that all three compounds amplified the accumulation of nuclear p53, elicited by benzo[a]pyrene (BaP) or dibenzo[al]pyrene (DBP). This effect was associated with an attenuated PAH-induced apoptosis and with decreased levels of phosphorylated FoxO3a Thr32. Thr32 phosphorylation of FoxO3a may promote a translocation of FoxO3a-p53 complex from nucleus to the cytoplasm, and the role of FoxO3a dephosphorylation was further studied. We found that inhibition of PP2A phosphatase restored levels of phosphorylated FoxO3a, led to cytosolic translocation of p53, and activated BaP-induced p53-mediated apoptosis. These results were confirmed by silencing FoxO3a with siRNA or by inhibiting 14-3-3 protein; also these treatments trapped BaP-induced p53 in the nucleus. Our data indicate interplay between p53, FoxO3a and 14-3-3 leading to an attenuated BaP induced apoptosis in cells co-exposed to TCDD, PCB 153 or estradiol.
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Supelco
Benzo[a]pyrene solution, 100 μg/mL in cyclohexane, analytical standard
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Supelco
Cycloheximide, PESTANAL®, analytical standard
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Dibutyl phthalate, European Pharmacopoeia (EP) Reference Standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 1000 μg/mL in acetone
Supelco
Dibutyl phthalate, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Supelco
Dibutyl phthalate, PESTANAL®, analytical standard
Supelco
Benzo[a]pyrene solution, certified reference material, TraceCERT®, 200 μg/mL in methylene chloride
Sigma-Aldrich
L-Glutamine
Supelco
Benzo[a]pyrene, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Supelco
Benzo[a]pyrene, analytical standard, for environmental analysis
Millipore
Cycloheximide solution, 0.1%, suitable for microbiology
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Cycloheximide solution, Ready-Made Solution, microbial, 100 mg/mL in DMSO, Suitable for cell culture
Sigma-Aldrich
Benzo[a]pyrene, ≥96% (HPLC)
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)