Merck
Interactions between phospholipids and titanium dioxide particles.
Colloids and surfaces. B, Biointerfaces (2014-09-23)
Quoc-Chon Le, Marie-Hélène Ropers, Hélène Terrisse, Bernard Humbert
ABSTRACT
A systematic study was carried out on monolayer films and lipid vesicles to elucidate the interactions between membrane lipids and commercial particles of titanium dioxide TiO2 (TiO2-P25). Pressure-area isotherms of lipids at various pH values were recorded on a Langmuir trough with or without TiO2-P25 and NaCl in the subphase. Electrophoretic mobilities of lipid vesicles and TiO2-P25 particles were measured to identify the pH range where attractive electrostatic interactions between lipids and TiO2-P25 could take place. The results show that (i) the surface of TiO2-P25 particles interacts only with some phospholipids, (ii) the driving forces are electrostatic and (iii) non-electrostatic interactions were also observed, depending on the molecular structure. More precisely, the phospholipids 1,2-dimyristoyl-sn-glycero-3-phosphate monosodium salt (DMPA), 1,2-dimyristoyl-sn-glycero-3-phospho-rac-1-glycerol (DMPG) and 1',3'-bis[1,2-dimyristoyl-sn-glycero-3-phospho]-sn-glycerol (TMCL) interacted strongly with the TiO2-P25 surface through electrostatic interactions, providing they were oppositely charged, i.e. for pH between 2 and 6.6. For TMCL and DMPG, interactions with the surface of TiO2-P25 in non-favourable electrostatic conditions, suggested another kind of binding, probably through the hydroxyl groups of the terminal glycerol. Weaker attractive interactions were demonstrated for 1,2-dimyristoyl-sn-glycero-3-phospho-l-serine (DMPS) and the synthetic lipid dihexadecyl phosphate (DHP). For DMPS, the carboxylate group is involved in the adsorption onto TiO2. The other membrane lipids such as 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine (DMPE) and sphingomyelin (SM) did not interact with TiO2-P25 regardless of pH.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3,4-Dihydro-2H-pyran, 97%
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Supelco
Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%