Merck
  • L-Ornithine phenylacetate reduces ammonia in pigs with acute liver failure through phenylacetylglycine formation: a novel ammonia-lowering pathway.
L-Ornithine phenylacetate reduces ammonia in pigs with acute liver failure through phenylacetylglycine formation: a novel ammonia-lowering pathway.
American journal of physiology. Gastrointestinal and liver physiology (2014-09-27)
Rune Gangsøy Kristiansen, Christopher F Rose, Ole-Martin Fuskevåg, Hanne Mæhre, Arthur Revhaug, Rajiv Jalan, Lars Marius Ytrebø
ABSTRACT
Glycine is an important ammoniagenic amino acid, which is increased in acute liver failure (ALF). We have previously shown that L-ornithine phenylacetate (OP) attenuates ammonia rise and intracranial pressure in pigs suffering from ALF but failed to demonstrate a stoichiometric relationship between change in plasma ammonia levels and excretion of phenylacetylglutamine in urine. The aim was to investigate the impact of OP treatment on the phenylacetylglycine pathway as an alternative and additional ammonia-lowering pathway. A well-validated and -characterized large porcine model of ALF (portacaval anastomosis, followed by hepatic artery ligation), which recapitulates the cardinal features of human ALF, was used. Twenty-four female pigs were randomized into three groups: (1) sham operated + vehicle, (2) ALF + vehicle, and (3) ALF + OP. There was a significant increase in arterial glycine concentration in ALF (P < 0.001 compared with sham), with a three-fold increase in glycine release into the systemic circulation from the kidney compared with the sham group. This increase was attenuated in both the blood and brain of the OP-treated animals (P < 0.001 and P < 0.05, respectively), and the attenuation was associated with renal removal of glycine through excretion of the conjugation product phenylacetylglycine in urine (ALF + vehicle: 1,060 ± 106 μmol/l; ALF + OP: 27,625 ± 2,670 μmol/l; P < 0.003). Data from this study provide solid evidence for the existence of a novel, additional pathway for ammonia removal in ALF, involving glycine production and removal, which is targeted by OP.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hippuric acid, 98%
Supelco
tert-Butyl methyl ether, analytical standard
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
SAFC
Glycine
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Alanine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
tert-Butyl methyl ether, ACS reagent, ≥99.0%
Supelco
tert-Butyl Methyl Ether, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
tert-Butyl methyl ether, reagent grade, ≥98%
Glycine, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
tert-Butyl methyl ether, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
tert-Butyl methyl ether, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Deuterium hydride, extent of labeling: 96 mol% DH, 98 atom % D