Merck
  • Effect of betamethasone in combination with antibiotics on gram positive and gram negative bacteria.
Effect of betamethasone in combination with antibiotics on gram positive and gram negative bacteria.
International journal of immunopathology and pharmacology (2015-01-13)
M Artini, R Papa, A Cellini, M Tilotta, G Barbato, A Koverech, L Selan
ABSTRACT
Betamethasone is an anti-inflammatory steroid drug used in cases of anaphylactic and allergic reactions, of Alzheimer and Addison diseases and in soft tissue injuries. It modulates gene expression for anti-inflammatory activity suppressing the immune system response. This latter effect might decrease the effectiveness of immune system response against microbial infections. Corticosteroids, in fact, mask some symptoms of infection and during their use superimposed infections may occur. Thus, the use of glucocorticoids in patients with sepsis remains extremely controversial. In this study we analyzed the in vitro effect of a commercial formulation of betamethasone (Bentelan) on several Gram positive and Gram negative bacteria of clinical relevance. It was found to be an inhibitor of the growth of most of the strains examined. Also the effect of betamethasone in combination with some classes of antibiotics was evaluated. Antibiotic-steroid combination therapy is, in such cases, superior to antibiotic-alone treatment to impair bacterial growths. Such effect was essentially not at all observable on Staphylococcus aureus or Coagulase Negative Staphylococci (CoNS).
Product Description
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Erythromycin, meets USP testing specifications
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
Ceftazidime, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Betamethasone, ≥98%
Ofloxacin, European Pharmacopoeia (EP) Reference Standard
Supelco
Betamethasone, VETRANAL®, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Ofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Betamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Betamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Ofloxacin, United States Pharmacopeia (USP) Reference Standard
USP
Betamethasone, United States Pharmacopeia (USP) Reference Standard