Merck
  • Plasma-based ambient sampling/ionization/transmission integrated source for mass spectrometry.
Plasma-based ambient sampling/ionization/transmission integrated source for mass spectrometry.
The Analyst (2014-08-26)
Yueming Zhou, Ning Zhang, Yafeng Li, Caiqiao Xiong, Suming Chen, Yongtai Chen, Zongxiu Nie
ABSTRACT
Better sensitivity and interface of ambient sampling/ionization mass spectrometry remain a challenge. Herein, a novel, plasma-based, ambient sampling/ionization/transmission (PASIT) integrated source in a pin-to-funnel configuration was developed for the sensitive analysis of complex samples. With the funnel sleeve directly affected by direct-current discharge plasma, PASIT combines the ability to sample/ionize analyte molecules and then efficiently collect/transport charged mass species under atmospheric pressure and consequently shows an improved sensitivity. The integrated source enhances the signal intensity by more than 2 orders of magnitude compared with the previous pin-to-plate plasma source without significant background addition. A surface limit of detection (LOD) of 130 fmol mm-(2) (S/N = 3) has been achieved for clenbuterol on filter paper with an argon carrier gas. Demonstrated applications include the direct determination of active ingredients from drugs and symbolic compounds from natural plants and cholesterol from mouse brain tissue sections.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
USP
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Caffeine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Melamine, 99%
Supelco
Melamine, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Melamine, United States Pharmacopeia (USP) Reference Standard
Supelco
Melting point standard 235-237°C, analytical standard