Merck
  • KDM4B as a target for prostate cancer: structural analysis and selective inhibition by a novel inhibitor.
KDM4B as a target for prostate cancer: structural analysis and selective inhibition by a novel inhibitor.
Journal of medicinal chemistry (2014-06-28)
Chia-Han Chu, Ling-Yu Wang, Kai-Cheng Hsu, Chung-Chin Chen, Hsing-Hung Cheng, Szu-Min Wang, Chien-Ming Wu, Tsan-Jan Chen, Ling-Ting Li, Ruiwu Liu, Chiu-Lien Hung, Jing-Moon Yang, Hsing-Jien Kung, Wen-Ching Wang
ABSTRACT
The KDM4/JMJD2 Jumonji C-containing histone lysine demethylases (KDM4A-KDM4D), which selectively remove the methyl group(s) from tri/dimethylated lysine 9/36 of H3, modulate transcriptional activation and genome stability. The overexpression of KDM4A/KDM4B in prostate cancer and their association with androgen receptor suggest that KDM4A/KDM4B are potential progression factors for prostate cancer. Here, we report the crystal structure of the KDM4B·pyridine 2,4-dicarboxylic acid·H3K9me3 ternary complex, revealing the core active-site region and a selective K9/K36 site. A selective KDM4A/KDM4B inhibitor, 4, that occupies three subsites in the binding pocket is identified by virtual screening. Pharmacological and genetic inhibition of KDM4A/KDM4B significantly blocks the viability of cultured prostate cancer cells, which is accompanied by increased H3K9me3 staining and transcriptional silencing of growth-related genes. Significantly, a substantial portion of differentially expressed genes are AR-responsive, consistent with the roles of KDM4s as critical AR activators. Our results point to KDM4 as a useful therapeutic target and identify a new inhibitor scaffold.
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
KDM4A/KDM4B Inhibitor, NSC636819
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)