Merck
  • A comparative study on properties of micro and nanopapers produced from cellulose and cellulose nanofibres.
A comparative study on properties of micro and nanopapers produced from cellulose and cellulose nanofibres.
Carbohydrate polymers (2014-12-30)
A Mtibe, Linda Z Linganiso, Aji P Mathew, K Oksman, Maya J John, Rajesh D Anandjiwala
ABSTRACT
Cellulose nanocrystals (CNCs) and cellulose nanofibres (CNFs) were successfully extracted from cellulose obtained from maize stalk residues. A variety of techniques, such as Fourier transform infrared (FTIR) spectroscopy, scanning electron microscopy (SEM), X-ray diffraction (XRD) and thermogravimetric analysis (TGA) were used for characterization and the experimental results showed that lignin and hemicellulose were removed to a greater extent by following the chemical methods. Atomic force microscopy (AFM) results confirmed that the diameters of CNCs and CNFs were ranging from 3 to 7 nm and 4 to 10nm, respectively, with their lengths in micro scale. CNCs suspension showed a flow of birefringence, however, the same was not observed in the case of suspension containing CNFs. XRD analysis confirmed that CNCs had high crystallinity index in comparison to cellulose and CNFs. Nanopapers were prepared from CNCs and CNFs by solvent evaporation method. Micropapers were also prepared from cellulose pulp by the same technique. Nanopapers made from CNFs showed less transparency as compared to nanopapers produced from CNCs whereas high transparency as compared to micropaper. Nanopapers produced from CNFs provided superior mechanical properties as compared to both micropaper and nanopapers produced from CNCs. Also, nanopapers produced from CNFs were thermally more stable as compared to nanopapers produced from CNCs but thermally less stable as compared to micropapers.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Potassium hydroxide, tested according to Ph. Eur.
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Sodium chlorite, puriss. p.a., 80% (RT)
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets