Merck
  • Systematic screening of different surface modifiers for the production of physically stable nanosuspensions.
Systematic screening of different surface modifiers for the production of physically stable nanosuspensions.
Journal of pharmaceutical sciences (2015-01-30)
Maria L A D Lestari, Rainer H Müller, Jan P Möschwitzer
ABSTRACT
The role of a surface modifier is important in the formation of stable nanosuspensions. In this study, a simple and systematic screening method for selecting optimum surface modifiers was performed by utilizing a low-energy wet ball milling method. Nine surface modifiers from different classes with different stabilization mechanisms were applied on six different models of active pharmaceutical ingredients (API). Particle size analysis showed that at concentration five times higher than the critical micelle concentration, SDS and sodium cholate (anionic surfactant) showed the highest percent success to produce stable nanosuspensions with particle size smaller than 250 nm. Similar findings were also shown by poloxamer 188 (nonionic surfactant) and hydroxypropylmethylcellulose E5 (polymeric stabilizer) at concentration 1% (w/v) and 0.8% (w/v), respectively. In addition, combinations of anionic surfactant and nonionic surfactant as well as combinations of anionic surfactant and polymeric stabilizer showed high percent success in the formation of stable nanosuspensions. In general, no correlation can be found between the physicochemical characteristics of the model API (molecular weight, melting point, log P, pKa, and crystallinity) with its feasibility to be nanosized. The concentration and the principle of stabilization of surface modifier determine the formation of stable nanosuspensions.
Product Description
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
USP
Ibuprofen, United States Pharmacopeia (USP) Reference Standard
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard