Merck
  • Effect of polydopamine on the biomimetic mineralization of mussel-inspired calcium phosphate cement in vitro.
Effect of polydopamine on the biomimetic mineralization of mussel-inspired calcium phosphate cement in vitro.
Materials science & engineering. C, Materials for biological applications (2014-10-05)
Zongguang Liu, Shuxin Qu, Xiaotong Zheng, Xiong Xiong, Rong Fu, Kuangyun Tang, Zhendong Zhong, Jie Weng
ABSTRACT
Inspired by the excellent adhesive property of mussel adhesive protein, we added polydopamine (PDA) to calcium phosphate cement (PDA-CPC) to enhance its compressive strength previously. The mineralization and mechanism on PDA-CPC were investigated by soaking it in simulated body fluid in this study. The results indicated that PDA promoted the conversion of dicalcium phosphate dihydrate and α-tricalcium phosphate to hydroxyapatite (HA) in the early stage but inhibited this conversion subsequently. PDA promoted the rapid mineralization on PDA-CPC to form a layer of nanoscale calcium phosphate (CaP) whereas there was no CaP formation on the control-CPC after 1d of soaking. This layer of nanoscale CaP was similar to that of natural bone, which was always observed during soaking. X-ray photoelectron spectroscopy showed that the peak of CO of PDA existed in the newly formed CaP on PDA-CPC, indicating the co-precipitation of CaP with PDA. Furthermore, the newly formed CaP on PDA-CPC was HA confirmed by transmission electron microscopy, which the newly formed HA was in association with PDA. Therefore, PDA increased the capacity of mineralization of CPC and induced the formation of nanoscale bone-like apatite on PDA-CPC. Thus, this provides the feasible route for surface modification on CPC.
Product Description
USP
Calcium Carbonate (AS), United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
α-Tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium carbonate, ≥99.995% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%
Sigma-Aldrich
Dicyclopentadiene, contains BHT as stabilizer
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Calcium carbonate, ACS reagent, 99.95-100.05% dry basis
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
β-tri-Calcium phosphate, puriss. p.a., ≥98% β-phase basis (sintered Powder)