Merck
  • Kaposi sarcoma herpes virus latency associated nuclear antigen protein release the G2/M cell cycle blocks by modulating ATM/ATR mediated checkpoint pathway.
Kaposi sarcoma herpes virus latency associated nuclear antigen protein release the G2/M cell cycle blocks by modulating ATM/ATR mediated checkpoint pathway.
PloS one (2014-06-28)
Amit Kumar, Sushil Kumar Sahu, Suchitra Mohanty, Sudipta Chakrabarti, Santanu Maji, R Rajendra Reddy, Asutosh K Jha, Chandan Goswami, Chanakya N Kundu, Shanmugam Rajasubramaniam, Subhash C Verma, Tathagata Choudhuri
ABSTRACT
The Kaposi's sarcoma-associated herpesvirus infects the human population and maintains latency stage of viral life cycle in a variety of cell types including cells of epithelial, mesenchymal and endothelial origin. The establishment of latent infection by KSHV requires the expression of an unique repertoire of genes among which latency associated nuclear antigen (LANA) plays a critical role in the replication of the viral genome. LANA regulates the transcription of a number of viral and cellular genes essential for the survival of the virus in the host cell. The present study demonstrates the disruption of the host G2/M cell cycle checkpoint regulation as an associated function of LANA. DNA profile of LANA expressing human B-cells demonstrated the ability of this nuclear antigen in relieving the drug (Nocodazole) induced G2/M checkpoint arrest. Caffeine suppressed nocodazole induced G2/M arrest indicating involvement of the ATM/ATR. Notably, we have also shown the direct interaction of LANA with Chk2, the ATM/ATR signalling effector and is responsible for the release of the G2/M cell cycle block.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mmab
Sigma-Aldrich
MISSION® esiRNA, targeting human ATM
Sigma-Aldrich
MISSION® esiRNA, targeting human MMAB
Sigma-Aldrich
MISSION® esiRNA, targeting human ATR
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Chek2
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
MISSION® esiRNA, targeting human ANTXR1
Sigma-Aldrich
MISSION® esiRNA, targeting human CHEK2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Antxr1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atr