Merck
  • Chrysin, baicalein and galangin are indirect activators of the human constitutive androstane receptor (CAR).
Chrysin, baicalein and galangin are indirect activators of the human constitutive androstane receptor (CAR).
Toxicology letters (2015-01-28)
Alejandro Carazo Fernández, Tomas Smutny, Lucie Hyrsová, Karel Berka, Petr Pavek
ABSTRACT
The constitutive androstane receptor (CAR) is a crucial transcriptional regulator of key xenobiotic-metabolizing enzymes such as cytochrome P450 CYP3A4, CYP2C9 and CYP2B6. The flavonoids chrysin, baicalein and galangin have been reported to activate CAR and interfere with EGFR signaling. Nevertheless, it is not known if these flavonoids are direct CAR ligands or indirect phenobarbital-like CAR activators via the inhibition of epidermal growth factor receptor (EGFR) signaling. We analyze the interactions of chrysin, galangin and baicalein and its glycoside baicalin with human CAR. We have employed and validated methods that can study direct interaction with the CAR ligand binding pocket. Secondly, we determined if the compounds affect human EGFR signaling and interact with EGFR. Employing a TR-FRET coactivator assay with recombinant CAR or CAR assembly assay, a consistent activation of CAR with flavonoids and phenobarbital was not observed. It was determined, however, that galangin, chrysin, and baicalein may slightly repress EGFR-Tyr1068 autophosphorylation after EGF treatment, phosphorylation of downstream transcription factor ELK1 and stimulate EGFP-CAR nuclear translocation in primary human hepatocytes. These data suggest that flavonoids chrysin, galangin and baicalein are indirect human CAR activators. This study also demonstrates new approach how to test the direct CAR interaction with its ligands.
Sigma-Aldrich
Galangin, autophagy inducing flavonoid
Sigma-Aldrich
5α-Androstan-3β-ol, powder
Supelco
Chrysin, analytical standard
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Baicalin, 95%
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Chrysin, ≥96.5%
Sigma-Aldrich
Baicalein, 98%
Sigma-Aldrich
Clotrimazole
Sigma-Aldrich
CITCO, ≥98% (HPLC), solid
Supelco
Clotrimazole, Pharmaceutical Secondary Standard; Certified Reference Material
Clotrimazole for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Baicalin, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Clotrimazole, European Pharmacopoeia (EP) Reference Standard
USP
Clotrimazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture