Merck
  • Free radical scavenging activity and comparative metabolic profiling of in vitro cultured and field grown Withania somnifera roots.
Free radical scavenging activity and comparative metabolic profiling of in vitro cultured and field grown Withania somnifera roots.
PloS one (2015-04-16)
Kalaiselvi Senthil, Pankajavalli Thirugnanasambantham, Taek Joo Oh, So Hyun Kim, Hyung Kyoon Choi
ABSTRACT
Free radical scavenging activity (FRSA), total phenolic content (TPC), and total flavonoid content (TFC) of in vitro cultured and field grown Withania somnifera (Ashwagandha) roots were investigated. Withanolides analysis and comprehensive metabolic profiling between 100% methanol extracts of in vitro and field grown root tissues was performed using high performance thin layer chromatography (HPTLC) and gas chromatography-mass spectrometry (GC-MS), respectively. Significantly higher levels of FRSA, TPC, and TFC were observed in in-vitro cultured roots compared with field grown samples. In addition, 30 day-cultured in vitro root samples (1 MIR) exhibited a significantly higher FRSA (IC50 81.01 μg/mL), TPC (118.91 mg GAE/g), and TFC (32.68 mg CE/g) compared with those in 45 day-cultured samples (1.5 MIR). Total of 29 metabolites were identified in in vitro cultured and field grown roots by GC-MS analysis. The metabolites included alcohols, organic acids, purine, pyrimidine, sugars, and putrescine. Vanillic acid was only observed in the in vitro cultured root samples, and higher level of the vanillic acid was observed in 1 MIR when compared to 1.5 MIR. Therefore, it is suggested that 1 MIR might serve as an alternative to field grown roots for the development of medicinal and functional food products.
Sigma-Aldrich
Methoxyamine hydrochloride, 98%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Aluminum chloride, 99.99% trace metals basis
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.99% trace metals basis
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Supelco
Sulfuric acid, for the determination of nitrogen, ≥97.0%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Aluminum chloride, anhydrous, powder, 99.999% trace metals basis
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)