Merck
  • Growth medium alterations improve in vitro cold storage of pear germplasm.
Growth medium alterations improve in vitro cold storage of pear germplasm.
Cryo letters (2014-07-07)
I Kovalchuk, Z Zhumagulova, T Turdiev, B M Reed
ABSTRACT
Development of new fruit cultivars is dependent on genetic resource collections such as those at the Pomological Garden of the Institute of Horticulture and Viticulture near Almaty, Kazakhstan. The pear germplasm collection of the Pomological Garden contains 615 cultivars and three species. In vitro cold storage of the collection would provide additional security to the field collection. This study was designed to improve medium-term in vitro storage of pear germplasm. Shoots of seven pear cultivars (Pyrus communis L.) were stored in plastic five-section bags at 4 degree C and a 10-h photoperiod (7 μmol/m2/s). Treatments included medium with four carbohydrate sources (3% sucrose, 2% or 3% mannitol, or 2% sucrose + 2% mannitol) with 0.5 mg/l BAP and 0.1 mg/l IBA or without plant growth regulators (PGRs) and at three Murashige and Skoog (MS) nitrogen concentrations (100%, 50% or 25%). Pear shoots remained viable for 9 to 15 months without repropagation on the control MS medium with 3% sucrose without PGRs. There were significant impacts of cultivar and treatment on the duration of cold storage. Shoots of 'Mramornaya' remained viable (rating of ≥ 2) for 27 months with PGRs and 2% sucrose + 2% mannitol compared to 12 months for the PGR + 3% sucrose treatment. Talgarskaya Krasaviza stored for 18 months on 2% sucrose + 2% mannitol while all other treatments lasted only 6 to 9 months. Treatments with 0.5 or 1 mg/l abscisic acid (ABA) with 3% sucrose increased storage duration as did reducing the concentration of nitrogen in the medium to 25% without PGRs and with 3% sucrose.
Sigma-Aldrich
Indole-3-butyric acid, suitable for plant cell culture, BioReagent
Sigma-Aldrich
Indole-3-butyric acid, ≥98.0% (T)