Merck
  • Double-effective chitosan scaffold-PLGA nanoparticle system for brain tumour therapy: in vitro study.
Double-effective chitosan scaffold-PLGA nanoparticle system for brain tumour therapy: in vitro study.
Journal of microencapsulation (2014-06-26)
Ceren Kutlu, Anil S Çakmak, Menemşe Gümüşderelioğlu
ABSTRACT
The chitosan scaffold, which has both of anticancer and antivascularization effects, was developed for using in local therapy of brain tumours. This is why, poly-lactic-co-glycolic acid (50:50) nanoparticles (~200 nm) including an anticancer drug, 5-fluorouracil (5-FU), were prepared by emulsion-solvent evaporation method. Then, these nanoparticles and antivascularization agent, bevacizumab, were loaded into the scaffold during manufacturing by freeze-drying and embedding after freeze-drying, respectively. The idea behind this system is to destroy tumour tissue by releasing 5-FU and to prevent the proliferation of tumour cells by releasing bevacizumab. In addition, 3D scaffold can support healthy tissue formation in the tumourigenic region. In vitro effectiveness of this system was investigated on T98G human glioblastoma cell line and human umbilical vein endothelial cells. The results show that the chitosan scaffold containing 100 µg 5-FU and 100 µg bevacizumab has a potential to prevent the tumour formation in vitro conditions.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, suitable for histochemical demonstration of nitro blue tetrazolium reduction in neutrophils
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, ACS reagent, 99%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, puriss. p.a., reag. Ph. Eur., ≥99% (perchloric acid titration)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Didodecyldimethylammonium bromide, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
4-(Dimethylamino)benzaldehyde, 98%
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
4-(Dimethylamino)benzaldehyde solution, 10 g/L in isopropanol, for TLC derivatization