Merck
  • Molecular characterization of ongoing enzymatic reactions in raw garlic cloves using extractive electrospray ionization mass spectrometry.
Molecular characterization of ongoing enzymatic reactions in raw garlic cloves using extractive electrospray ionization mass spectrometry.
Analytical chemistry (2015-02-14)
Hua Zhang, Konstantin Chingin, Liang Zhu, Huanwen Chen
ABSTRACT
Characterization of enzymatic reactions occurring in untreated biological samples is of increasing interest. Herein, the chemical conversion of alliin to allicin, catalyzed by allinase, in raw garlic cloves has been followed in vivo by internal extractive electrospray ionization mass spectrometry (iEESI-MS). Both precursors and products of the enzymatic reaction were instantaneously extracted by infused solution running throughout the tissue and directly electrospray ionized on the edge of the bulk sample for online MS analysis. Compared to the room-temperature (+25 °C) scenario, the alliin conversion in garlic cloves decreased by (7.2 ± 1.4) times upon heating to +80 °C and by (5.9 ± 0.8) times upon cooling to -16 °C. Exposure of garlic to gentle ultrasound irradiation for 3 h accelerated the reaction by (1.2 ± 0.1) times. A 10 s microwave irradiation promoted alliin conversion by (1.6 ± 0.4) times, but longer exposure to microwave irradiation (90 s) slowed the reaction by (28.5 ± 7.5) times compared to the reference analysis. This method has been further employed to monitor the germination process of garlic. These data revealed that over a 2 day garlic sprouting, the allicin/alliin ratio increased by (2.2 ± 0.5) times, and the averaged degree of polymerization for the detected oligosaccharides/polysaccharides decreased from 11.6 to 9.4. Overall, these findings suggest the potential use of iEESI-MS for in vivo studies of enzymatic reactions in native biological matrices.
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Supelco
(+)-L-Alliin, analytical standard
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
Supelco
Sucrose, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sucrose, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
(±)-L-Alliin, ≥90% (HPLC)
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Millipore
Sucrose, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium hydroxide solution, BioUltra, ~1 M NH3 in H2O (T)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Sucrose, analytical standard, for enzymatic assay kit SCA20
Sucrose, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium hydroxide solution, ACS reagent, 28.0-30.0% NH3 basis