Merck
  • A multiclass method for the analysis of endocrine disrupting chemicals in human urine samples. Sample treatment by dispersive liquid-liquid microextraction.
A multiclass method for the analysis of endocrine disrupting chemicals in human urine samples. Sample treatment by dispersive liquid-liquid microextraction.
Talanta (2014-08-17)
F Vela-Soria, O Ballesteros, A Zafra-Gómez, L Ballesteros, A Navalón
ABSTRACT
The population is continuously exposed to endocrine disrupting chemicals (EDCs). This has influenced an increase in diseases and syndromes that are more frequent nowadays. Therefore, it is necessary to develop new analytical procedures to evaluate the exposure with the ultimate objective of establishing, in an accurate way, relationships between EDCs and harmful health effects. In the present work, a new method based on a sample treatment by dispersive liquid-liquid microextraction (DLLME) for the extraction of six parabens (methyl-, ethyl-, isopropyl-, propyl-, isobutyl and butylparaben), six benzophenones (benzophenone-1, benzophenone-2, benzophenone-3, benzophenone-6, benzophenone-8 and 4-hydroxybenzophenone) and two bisphenols (bisphenol A and bisphenol S) in human urine samples, followed by gas chromatography-tandem mass spectrometry (GC-MS/MS) analysis is proposed. An enzymatic treatment allows determining the total content of the target EDCs. The extraction parameters were accurately optimized using multivariate optimization strategies. Ethylparaben ring-(13)C6 and bisphenol A-d16 were used as surrogates. Found limits of quantification ranging from 0.2 to 0.5 ng mL(-1) and inter-day variability (evaluated as relative standard deviation) ranging from 2.0% to 14.9%. The method was validated using matrix-matched standard calibration followed by a recovery assay with spiked samples. Recovery rates ranged from 94% to 105%. A good linearity, for concentrations up to 300 ng mL(-1) for parabens and 40 ng mL(-1) for benzophenones and bisphenols, respectively, was obtained. The method was satisfactorily applied for the determination of target compounds in human urine samples from 20 randomly selected individuals.
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Chlorotrimethylsilane, puriss., ≥99.0% (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Potassium chloride, tested according to Ph. Eur.
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Supelco
Potassium chloride solution, BioUltra, ~3 M in H2O
Supelco
Carbon tetrachloride, analytical standard
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 1% TMCS, 99% (excluding TMCS)
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Lactic acid, natural, ≥85%
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, for GC derivatization, LiChropur, contains 10% TMCS, 98% (excluding TMCS)
Sigma-Aldrich
Calcium chloride dihydrate, BioUltra, for molecular biology, ≥99.5% (KT)
Supelco
Potassium chloride solution, for Ag/AgCl electrodes, ~3 M KCl, saturated with silver chloride
Sigma-Aldrich
Lactic acid, 85%, FCC