Merck
  • Using in situ Raman spectroscopy to study the drug precipitation inhibition and supersaturation mechanism of Vitamin E TPGS from self-emulsifying drug delivery systems (SEDDS).
Using in situ Raman spectroscopy to study the drug precipitation inhibition and supersaturation mechanism of Vitamin E TPGS from self-emulsifying drug delivery systems (SEDDS).
Journal of pharmaceutical and biomedical analysis (2015-03-15)
Shilpa Raut, Basel Karzuon, Eman Atef
ABSTRACT
We are reporting a new methodology of using Raman spectroscopy for studying the drug surfactant interactions in self-emulsifying drug delivery systems (SEDDS). The physicochemical properties of surfactants could affect the performance of drugs from lipid delivery systems. Thus the purpose of our research was to study the drug surfactant interactions on a molecular level to understand the mechanism of supersaturation and precipitation inhibition. Two surfactants, Labrasol® and Vitamin E TPGS, were used to formulate several SEDDS. The optimized SEDDS were further evaluated by a kinetic solubility study and in situ Raman spectroscopy for two model drugs. It was found that both drugs precipitated from Labrasol® SEDDS whereas TPGS was able to inhibit precipitation and achieve high drug supersaturation levels. In situ Raman spectroscopy indicated that hydrogen bonding with TPGS was the main factor responsible for inhibiting precipitation. This study was able to correlate the structure and physicochemical properties of the drugs and surfactants to their ability to prevent drug precipitation. Our study brings up a possible new systematic approach by using Raman spectroscopy in the development and optimization of lipid based delivery systems.
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Glibenclamide for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%