Merck
Effects of metformin on retinoblastoma growth in vitro and in vivo.
International journal of oncology (2014-09-13)
Katarzyna Brodowska, Sofia Theodoropoulou, Melissa Meyer Zu Hörste, Eleftherios I Paschalis, Kimio Takeuchi, Gordon Scott, David J Ramsey, Elizabeth Kiernan, Mien Hoang, Joanna Cichy, Joan W Miller, Evangelos S Gragoudas, Demetrios G Vavvas
ABSTRACT
Recent studies suggest that the anti-diabetic drug metformin may reduce the risk of cancer and have anti-proliferative effects for some but not all cancers. In this study, we examined the effects of metformin on human retinoblastoma cell proliferation in vitro and in vivo. Two different human retinoblastoma cell lines (Y79, WERI) were treated with metformin in vitro and xenografts of Y79 cells were established in nu/nu immune-deficient mice and used to assess the effects of pharmacological levels of metformin in vivo. Metformin inhibited proliferation of the retinoblastoma cells in vitro. Similar to other studies, high concentrations of metformin (mM) blocked the cell cycle in G0‑G1, indicated by a strong decrease of G1 cyclins, especially cyclin D, cyclin-dependent kinases (4 and 6), and flow cytometry assessment of the cell cycle. This was associated with activation of AMPK, inhibition of the mTOR pathways and autophagy marker LC3B. However, metformin failed to suppress growth of xenografted tumors of Y79 human retinoblastoma cells in nu/nu mice, even when treated with a maximally tolerated dose level achieved in human patients. In conclusion, suprapharmacological levels (mM) of metformin, well above those tolerated in vivo, inhibited the proliferation of retinoblastoma cells in vitro. However, physiological levels of metformin, such as seen in the clinical setting, did not affect the growth of retinoblastoma cells in vitro or in vivo. This suggests that the potential beneficial effects of metformin seen in epidemiological studies may be limited to specific tumor types or be related to indirect effects/mechanisms not observed under acute laboratory conditions.
Sigma-Aldrich
MOPS, BioUltra, for molecular biology, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
SAFC
HEPES
SAFC
BIS-TRIS
SAFC
BIS-TRIS
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
SAFC
HEPES
SAFC
MOPS
Sigma-Aldrich
Mouse IGF-I ELISA Kit, for serum, plasma and cell culture supernatant
Sigma-Aldrich
Human IGF-I R ELISA Kit, for serum, plasma, cell culture supernatant and urine
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Calcein, Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Human IGF-I ELISA Kit, for serum, plasma, cell culture supernatant and urine