Merck
  • Reversed expression of GRIM-1 and GRP78 in human non-small cell lung cancer.
Reversed expression of GRIM-1 and GRP78 in human non-small cell lung cancer.
Human pathology (2014-08-02)
Hui-Mei Wu, Zi-Feng Jiang, Xiao-Yun Fan, Tong Wang, Ke-Xu, Xue-Bo Yan, Yang Ma, Wei-Hua Xiao, Rong-Yu Liu
ABSTRACT
Gene associated with retinoid and interferon-induced mortality 1 (GRIM-1) acts as a tumor growth suppressor via apoptosis induction. However, GRIM-1 expression in human non-small cell lung cancer (NSCLC) and its potential interaction with another apoptosis-associated protein-glucose-regulated protein 78 (GRP78)-are as yet unknown. Using 40 surgical specimens, we showed significantly lower expression of GRIM-1 in NSCLC at both protein and messenger RNA (mRNA) levels compared with that in normal tissues (P < .01 and P < .001, respectively). Interestingly, these tumors tended to express higher basal amounts of GRP78 protein and mRNA (P < .05 and P < .001, respectively). Similarly, in the NSCLC tissues, weaker staining for GRIM-1 (main intensity + to ++) but stronger staining for GRP78 (main intensity +++ to ++++) was observed. Correlation analysis showed that protein and mRNA expression or the percentage of cells immunoreactive for GRIM-1 was negatively correlated with that of GRP78 (r = -0.279, r = -0.326, or r = -0.571, respectively). Coimmunoprecipitation and transient transfection revealed that GRIM-1 interacted with GRP78 and suppressed GRP78 protein expression. In addition, there was no correlation between GRIM-1 expression and clinical characteristics, whereas GRP78 expression was significantly correlated with tumor-nodes-metastasis (TNM) stage (stage 3 + 4 versus stage 1 + 2). In conclusion, the expression of GRIM-1 and GRP78 was negatively correlated in human NSCLC tissues, and the down-regulation of GRP78 by GRIM-1 provides a possible mechanism for their interaction. This study suggests a novel potential molecular pathway inactivated during the development of NSCLC.
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard