Merck
  • Evaluation of clinical and cytogenetic parameters in rheumatoid arthritis patients for effective diagnosis.
Evaluation of clinical and cytogenetic parameters in rheumatoid arthritis patients for effective diagnosis.
Clinica chimica acta; international journal of clinical chemistry (2014-10-14)
R Chandirasekar, B Lakshman Kumar, R Jayakumar, V Uthayakumar, Raichel Jacob, K Sasikala
ABSTRACT
Rheumatoid arthritis is the commonest inflammatory joint disease, affecting nearly 1% of the adult population worldwide. Early and accurate diagnosis and prognosis of rheumatoid arthritis (RA) have become increasingly important. In the present study, we aimed to elucidate the relationships between hematological, biochemical, immunological and cytogenetic parameters in rheumatoid arthritis patients and healthy normal controls. The study group comprised of 126 RA patients and equal number of healthy normal control subjects. The blood was collected and analyzed for biochemical, immunological, enzymatic and cytogenetic parameters. Results of the present study indicated that 20% of RA patient's hematological, 31% of biochemical and 70% immunological parameters had a significant difference from the controls and reference range. The RF and anti-CCP antibody levels were also positive in 70% of RA patients. A significant increase in minor chromosomal abnormalities was also observed in patients as compared to controls. The knowledge about autoimmune diseases is very low among the South Indian population. The present study has thus helped in understanding the RA disease in a better way based on a pattern of various clinical markers of the disease condition which might help in planning therapeutic intervention strategies and create awareness about the disease management among RA patients of the population studied.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))