Merck
  • Development of a multi-residue method in a fetal matrix: analysis of meconium.
Development of a multi-residue method in a fetal matrix: analysis of meconium.
Analytical and bioanalytical chemistry (2014-11-10)
Marie Meyer-Monath, Claudine Chatellier, Florence Rouget, Isabelle Morel, Francois Lestremau
ABSTRACT
Meconium is the earliest stool of newborns. It is a complex matrix that reflects the degree of fetal exposure to environmental pollutants. To investigate exposure to xenobiotics, an analytical method was developed to identify and quantify some pesticides and their metabolites and BTEX metabolites in meconium. Samples were prepared by two liquid-solid extractions and purified twice using SPE cartridges, followed by analysis with liquid chromatography coupled with tandem mass spectrometry. SPE cartridges (polymeric phase with hydrophilic and hydrophobic interactions, ion exchange, mixed mode) were tested and matrix effects were evaluated to determine purification performance. The quantification limits in meconium of this multi-residue method were in the range of 30 ng g(-1). The analytical method was applied to "real" meconium samples. Some target analytes were determined in most samples.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Mandelic acid, 99%
Supelco
o-Cresol, analytical standard
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Sodium sulfate, tested according to Ph. Eur., anhydrous
Supelco
β-Cyfluthrin, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Phenylglyoxylic acid, 97%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Supelco
α-Cypermethrin, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Supelco
Dichlorvos, PESTANAL®, analytical standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
o-Cresol, ≥99%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Supelco
Propoxur, PESTANAL®, analytical standard