Merck
  • Differential modulation of donor-specific antibodies after B-cell depleting therapies to cure chronic antibody mediated rejection.
Differential modulation of donor-specific antibodies after B-cell depleting therapies to cure chronic antibody mediated rejection.
Transplantation (2014-07-17)
Maxime Touzot, Grégoire Couvrat-Desvergnes, Stéphanie Castagnet, Anne Cesbron, Karine Renaudin, Diego Cantarovich, Magali Giral
ABSTRACT
Donor-specific antibodies (DSA) are considered as reliable biomarkers for antibody-mediated rejection (ABMR) diagnosis. However, it is unclear whether DSA monitoring is necessary and could predict graft outcome after antirejection treatment. We analyzed 28 non-sensitized kidney transplant patients with ABMR associated with de novo anti-human leukocyte antigen (HLA) DSA. Donor-specific antibody levels were measured by single antigen bead assays 12 months after antirejection therapy onset. Patients were placed in three groups according to their antirejection treatment: group I (n = 10), plasma exchange-Rituximab; group II (n = 8), Bortezomib; and group III (n = 10), optimization of maintenance immunosuppression. Half of the patients in group I demonstrated concomitant acute cellular rejection (ACR+). De novo DSA were mainly anti-DQ (60%). Anti-class I and anti-DR DSA disappeared after treatment in group I and remained negative during follow-up, whereas anti-DQ DSA persisted without any modulation. In contrast, class I-II HLA-DSA mean fluorescence intensity remained unchanged in groups II and III.Graft loss was observed in 80% and 20% of patients from group I (ACR+) and group III, respectively. One year after the ABMR treatment, a 16-mL/min decline in estimated glomerular filtration rate was observed in patients from group I (ACR-) and group III. Group II showed better outcomes with a mean estimated glomerular filtration rate decline of 6.4 mL/min. Modulation of DSA at and after treatment of ABMR did not correlate with graft outcome over a 12-month period.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Prednisone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prednisone, ≥98%
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard