Merck
  • Total synthesis and antiviral activity of indolosesquiterpenoids from the xiamycin and oridamycin families.
Total synthesis and antiviral activity of indolosesquiterpenoids from the xiamycin and oridamycin families.
Nature communications (2015-02-05)
Zhanchao Meng, Haixin Yu, Li Li, Wanyin Tao, Hao Chen, Ming Wan, Peng Yang, David J Edmonds, Jin Zhong, Ang Li
ABSTRACT
Indolosesquiterpenoids are a growing class of natural products that exhibit a wide range of biological activities. Here, we report the total syntheses of xiamycin A and oridamycins A and B, indolosesquiterpenoids isolated from Streptomyces. Two parallel strategies were exploited to forge the carbazole core: 6π-electrocyclization/aromatization and indole C2-H bond activation/Heck annulation. The construction of their trans-decalin motifs relied on two diastereochemically complementary radical cyclization reactions mediated by Ti(III) and Mn(III), respectively. The C23 hydroxyl of oridamycin B was introduced by an sp(3) C-H bond oxidation at a late stage. On the basis of the chemistry developed, the dimeric congener dixiamycin C has been synthesized for the first time. Evaluation of the antiviral activity of these compounds revealed that xiamycin A is a potent agent against herpes simplex virus-1 (HSV-1) in vitro.
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Potassium permanganate, ≤150 μm particle size, 97%
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Potassium permanganate, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Ammonium molybdate tetrahydrate, ACS reagent, 99.98% trace metals basis