Merck
  • Modulation of heparan sulfate in the glomerular endothelial glycocalyx decreases leukocyte influx during experimental glomerulonephritis.
Modulation of heparan sulfate in the glomerular endothelial glycocalyx decreases leukocyte influx during experimental glomerulonephritis.
Kidney international (2014-04-25)
Angelique L W M M Rops, Markus A Loeven, Jasper J van Gemst, Iris Eversen, Xander M Van Wijk, Henry B Dijkman, Toin H van Kuppevelt, Jo H M Berden, Ton J Rabelink, Jeffrey D Esko, Johan van der Vlag
ABSTRACT
The glomerular endothelial glycocalyx is postulated to be an important modulator of permeability and inflammation. The glycocalyx consists of complex polysaccharides, the main functional constituent of which, heparan sulfate (HS), is synthesized and modified by multiple enzymes. The N-deacetylase-N-sulfotransferase (Ndst) enzymes initiate and dictate the modification process. Here we evaluated the effects of modulation of HS in the endothelial glycocalyx on albuminuria and glomerular leukocyte influx using mice deficient in endothelial and leukocyte Ndst1 (TEKCre+/Ndst1flox/flox). In these mice, glomerular expression of a specific HS domain was significantly decreased, whereas the expression of other HS domains was normal. In the endothelial glycocalyx, this specific HS structure was not associated with albuminuria or with changes in renal function. However, glomerular leukocyte influx was significantly reduced during antiglomerular basement membrane nephritis, which was associated with less glomerular injury and better renal function. In vitro decreased adhesion of wild-type and Ndst1-deficient granulocytes to Ndst1-silenced glomerular endothelial cells was found, accompanied by a decreased binding of chemokines and L-selectin. Thus, modulation of HS in the glomerular endothelial glycocalyx significantly reduced the inflammatory response in antiglomerular basement membrane nephritis.
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sigma-Aldrich
ANTI-NIK (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material