Merck
  • Preclinical combination therapy of the investigational drug NAMI-A(+) with doxorubicin for mammary cancer.
Preclinical combination therapy of the investigational drug NAMI-A(+) with doxorubicin for mammary cancer.
Investigational new drugs (2014-10-24)
Alberta Bergamo, Tina Riedel, Paul J Dyson, Gianni Sava
ABSTRACT
The tumor metastases targeting ruthenium complex NAMI-A synergistically improves the activity of gemcitabine in combination therapies. High-throughput screening was used to identify other potential drug combinations from a library of FDA approved drugs. Doxorubicin was identified as a hit compound and was therefore evaluated in combination with NAMI-A in vitro and in a preclinical in vivo model. High-throughput screening identified eight structurally diverse compounds that synergize with NAMI-A including doxorubicin. The combination index on MCF-7 cells showed synergism as the concentration of NAMI-A increases independent of the doxorubicin concentration. In MCa mammary carcinoma of CBA mice, NAMI-A (35 mg/kg/day i.p. on days 7-12) followed by doxorubicin (10 mg/kg i.p. on day 16), significantly increased the effects of the individual drugs on metastases with 70 % animals resulting free of macroscopically detectable tumor nodules in the lungs at sacrifice. NAMI-A, unlike doxorubicin, cured 60 % of the treated mice but the combination therapy was toxic to the animals. The combined therapy of NAMI-A with doxorubicin synergizes on lung metastasis in a preclinical mouse model. The combination therapy at the maximum tolerated doses of the two drugs is toxic. Hence, this combination is not suitable for clinical studies using maximum tolerated doses.
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Streptomycin sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Glutamine
Supelco
Streptomycin solution, ~1 mg/mL in 1 mM EDTA, analytical standard
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Streptomycin Ready Made Solution, 100 mg/mL in water