Merck
  • Hapten synthesis and development of a competitive indirect enzyme-linked immunosorbent assay for acrylamide in food samples.
Hapten synthesis and development of a competitive indirect enzyme-linked immunosorbent assay for acrylamide in food samples.
Journal of agricultural and food chemistry (2014-07-08)
Jing Wu, Yu-Dong Shen, Hong-Tao Lei, Yuan-Ming Sun, Jin-Yi Yang, Zhi-Li Xiao, Hong Wang, Zhen-Lin Xu
ABSTRACT
The high level of acrylamide in widely consumed processed foods poses a potentially significant risk to human health, which has led to an increasing demand for rapid, simple, and selective analytical methods. In the present work, several haptens for acrylamide were designed in an attempt to prepare antibodies with acrylamide affinity, but they failed their purpose. However, a polyclonal antibody was produced against 4-mercaptophenylacetic acid (4-MPA)-derivatized acrylamide, which showed high binding affinity to the derivative. As acrylamide easily reacted with 4-MPA at high derivation yield, a competitive indirect enzyme-linked immunosorbent assay (ciELISA) for acrylamide via a preanalysis derivatization was developed. The derivatization and ELISA conditions were fully optimized to produce a method for acrylamide assay that exhibited an IC50 of 2.86 μg/kg, limit of detection at 0.036 μg/kg, and linear range of 0.25-24.15 μg/kg. The results of preanalysis recovery tests of acrylamide-spiked food samples and screening of blind food samples by both ciELISA and HPLC-MS/MS indicated the proposed ciELISA's good accuracy and reliability. This method was thus deemed suitable for routine acrylamide screening in food samples at low cost.
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Acrylamide, for molecular biology, ≥99% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
Maleamic acid
Sigma-Aldrich
Methyl acrylate, 99%, contains ≤100 ppm monomethyl ether hydroquinone as inhibitor
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylic acid, anhydrous, contains 200 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Supelco
Methyl acrylate, analytical standard
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Methacrylamide, 98%
Sigma-Aldrich
DL-2-Aminobutyric acid, ReagentPlus®, 99%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
N-Hydroxysuccinimide, purum, ≥97.0% (T)
Supelco
Acrylamide solution, 40% in H2O, for molecular biology
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered