Merck
  • Sulfide oxidations for LC-MS analysis of methionine-containing microcystins in Dolichospermum flos-aquae NIVA-CYA 656.
Sulfide oxidations for LC-MS analysis of methionine-containing microcystins in Dolichospermum flos-aquae NIVA-CYA 656.
Environmental science & technology (2014-10-22)
Christopher O Miles, Jeremy E Melanson, Andreas Ballot
ABSTRACT
Microcystins are cyclic heptapeptides produced by a range of cyanobacteria. More than 150 microcystin analogues have been reported from cultures, algal blooms, or other contaminated samples. Relatively few analytical standards are available, making identification and quantitation of these toxins a challenge, even with LC-MS technology. We developed a two-step oxidative procedure that allows LC-MS identification of microcystins containing methionine and methionine sulfoxide, and reveals the oxidation state of the methionyl sulfur atom. The procedure was used in parallel with mercaptoethanol derivatization and LC-MS(2) analysis to demonstrate the presence of [Asp(3)]MC-MR (12) and MC-MR (17) in a culture of Dolichospermum flos-aquae, together with low levels of [Asp(3)]MC-M(O)R (5) and MC-M(O)R (7), as well as 20 other microcystins. Fresh culture contained only traces of sulfoxides 5 and 7, but these increased during storage or sample extraction and preparation. This suggests that microcystins containing methionine sulfoxide are primarily postextraction oxidation artifacts, rather than being produced by biosynthesis in cyanobacteria. A simple, rapid extraction under inert gas followed promptly by LC-MS analysis minimized oxidation artifacts for D. flos-aquae.
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Supelco
2-Mercaptoethanol, for HPLC derivatization, LiChropur, ≥99.0% (GC)
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, meets USP testing specifications
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 30 wt. % in H2O, ACS reagent
Sigma-Aldrich
Hydrogen peroxide solution, 50 wt. % in H2O, stabilized
Sigma-Aldrich
Hydrogen peroxide solution, contains inhibitor, 35 wt. % in H2O
Sigma-Aldrich
OXONE®, monopersulfate compound
Sigma-Aldrich
OXONE®, monopersulfate compound, free-flowing, Redi-Dri