Merck
  • GC-MS, LC-MS(n), LC-high resolution-MS(n), and NMR studies on the metabolism and toxicological detection of mesembrine and mesembrenone, the main alkaloids of the legal high "Kanna" isolated from Sceletium tortuosum.
GC-MS, LC-MS(n), LC-high resolution-MS(n), and NMR studies on the metabolism and toxicological detection of mesembrine and mesembrenone, the main alkaloids of the legal high "Kanna" isolated from Sceletium tortuosum.
Analytical and bioanalytical chemistry (2014-09-23)
Golo M J Meyer, Carina S D Wink, Josef Zapp, Hans H Maurer
ABSTRACT
Mesembrine and mesembrenone are the main alkaloids of Sceletium tortuosum, a plant species that was used for sedation and analgesia by the KhoiSan, previously known as Hottentots, a tribe in South Africa. After fermentation, the obtained preparation called "Kanna" or "Kougoed" was used by chewing, smoking, or sniffing. Today, Kanna gains popularity by drug users as legal high. For monitoring such consumption, metabolism studies are mandatory because the metabolites are mostly the analytical targets, especially in urine. Therefore, the metabolism of both alkaloids was investigated in rat urine and pooled human liver preparations after several sample work-up procedures. As both alkaloids were not commercially available, they were isolated from plant material by Soxhlet extraction, and their identity confirmed by NMR. The metabolites were identified using gas chromatography-mass spectrometry (GC-MS) and liquid chromatography coupled to linear ion trap high resolution mass spectrometry (LC-HR-MS(n)). Both alkaloids were O- and N-demethylated, dihydrated, and/or hydroxylated at different positions. The phenolic metabolites were partly excreted as glucuronides and/or sulfates. Most of the phase I metabolites identified in rat urine could be detected also in the human liver preparations. After a common user's low dose application of mesembrine, mainly the O- and N demethyl-dihydro, hydroxy, and bis-demethyl-dihydro metabolites, and in case of mesembrenone only the N-demethyl and the N-demethyl-dihydro metabolite could be detected in rat urine using the authors' standard urine screening approaches (SUSA) by GC-MS or LC-MS(n). Thus, it should be possible to monitor a consumption of mesembrine and/or mesembrenone assuming similar pharmacokinetics in humans.
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Calcium formate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Calcium formate, BioUltra, ≥99.0% (T)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), for GC derivatization, LiChropur
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Aluminum oxide, mesoporous, MSU-X (wormhole), average pore size 3.8 nm
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Diethyl ether, analytical standard
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), for GC derivatization, LiChropur
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D